Insulin (Pork)
Synonym(s):
Insulin from porcine pancreas, Insulin from hog pancreas
Empirical Formula (Hill Notation):
C256H381N65O76S6
CAS Number:
Molecular Weight:
5777.54
Products
Product Category
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing