Skip to Content
MilliporeSigma
vibrant-m

Insulin lispro

vibrant-m
Empirical Formula (Hill Notation):
C257H383N65O77S6
CAS Number:
Molecular Weight:
5807.57

Products

Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade