Skip to Content
MilliporeSigma
Inosine ≥99% (HPLC)

Inosine

Inosine ≥99% (HPLC)
Synonym(s):
(−)-Inosine, Hypoxanthine 9-β-D-ribofuranoside, Inosine
Empirical Formula (Hill Notation):
C10H12N4O5
CAS Number:
Molecular Weight:
268.23

Products

Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment