Skip to Content
MilliporeSigma
Indoxyl β-D-glucoside ≥97%

Indoxyl β-D-glucoside

Indoxyl β-D-glucoside ≥97%
Synonym(s):
3-(β-D-Glucosido)indole, Plant indican
Empirical Formula (Hill Notation):
C14H17NO6
CAS Number:
Molecular Weight:
295.29

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing