Skip to Content
MilliporeSigma
Indium(III) trifluoromethanesulfonate

Indium(III) trifluoromethanesulfonate

Indium(III) trifluoromethanesulfonate
Synonym(s):
In(TFA)3, Indium(III) triflate, Trifluoromethanesulfonic acid indium(III) salt, Tris(trifluoromethanesulfonato)indium
Linear Formula:
(CF3SO3)3In
CAS Number:
Molecular Weight:
562.03

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing