Skip to Content
MilliporeSigma
Indigotindisulfonate sodium United States Pharmacopeia (USP) Reference Standard

Indigotindisulfonate sodium

Indigotindisulfonate sodium United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Indigo carmine, Acid Blue 74, Indigo-5,5′-disulfonic acid disodium salt, Indigocarmine
Empirical Formula (Hill Notation):
C16H8N2Na2O8S2
CAS Number:
Molecular Weight:
466.35

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1