Skip to Content
MilliporeSigma
Imipramine hydrochloride British Pharmacopoeia (BP) Reference Standard

Imipramine hydrochloride

Imipramine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
10,11-Dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, 5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenz[b,f]azepine hydrochloride, Imipramine hydrochloride
Empirical Formula (Hill Notation):
C19H24N2 · HCl
CAS Number:
Molecular Weight:
316.87
EC Number:

Products

Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique