Skip to Content
MilliporeSigma
Iminodi(methylphosphonic acid) 97%

Iminodi(methylphosphonic acid)

Iminodi(methylphosphonic acid) 97%
Linear Formula:
HN[CH2PO(OH)2]2
CAS Number:
Molecular Weight:
205.04

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing