Skip to Content
MilliporeSigma
Imidazolidinyl urea

Imidazolidinyl urea

Imidazolidinyl urea
Synonym(s):
N,N′′-Methylenebis[N′-[3-(hydroxymethyl)-2,5-dioxo-4-imidazolidinyl]-urea
Empirical Formula (Hill Notation):
C11H16N8O8
CAS Number:
Molecular Weight:
388.29
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing