Skip to Content
MilliporeSigma
Imazalil certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Imazalil

Imazalil certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1-[2-(Allyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole
Empirical Formula (Hill Notation):
C14H14Cl2N2O
CAS Number:
Molecular Weight:
297.18

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique