Skip to Content
MilliporeSigma
Iloperidone ≥98% (HPLC)

Iloperidone

Iloperidone ≥98% (HPLC)
Synonym(s):
1-[4-[3-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]propoxy]-3-methoxyphenyl]ethanone, Zomaril
Empirical Formula (Hill Notation):
C24H27FN2O4
CAS Number:
Molecular Weight:
426.48

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing