Skip to Content
MilliporeSigma
ICMA 97% (HPLC)

ICMA

ICMA 97% (HPLC)
Synonym(s):
1′,4′-Dihydro-naphtho[2′,3′:1,2][5,6]fullerene-C60, C60 derivative, indene-C60 monoadduct
Empirical Formula (Hill Notation):
C69H8
CAS Number:
Molecular Weight:
836.80

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing