Skip to Content
MilliporeSigma
Hyodeoxycholic acid methyl ester ≥98%

Hyodeoxycholic acid methyl ester

Hyodeoxycholic acid methyl ester ≥98%
Synonym(s):
3α,6α-Dihydroxy-5β-cholan-24-oic acid methyl ester
Empirical Formula (Hill Notation):
C25H42O4
CAS Number:
Molecular Weight:
406.60

Products

Product Category
Brand
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing