Skip to Content
MilliporeSigma
Hydroxypropyl acrylate, mixture of isomers contains 200 ppm hydroquinone monomethyl ether as inhibitor, 95%

Hydroxypropyl acrylate, mixture of isomers

Hydroxypropyl acrylate, mixture of isomers contains 200 ppm hydroquinone monomethyl ether as inhibitor, 95%
Linear Formula:
H2C=CHCO2C3H6OH
CAS Number:
Molecular Weight:
130.14
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type