Skip to Content
MilliporeSigma
Hydroxyectoine ≥95% (HPLC)

Hydroxyectoine

Hydroxyectoine ≥95% (HPLC)
Synonym(s):
(4S,5S)-5-Hydroxy-2-methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic acid, THP(A)
Empirical Formula (Hill Notation):
C6H10N2O3
CAS Number:
Molecular Weight:
158.16

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing