Skip to Content
MilliporeSigma
Hydrogen ionophore II Selectophore™

Hydrogen ionophore II

Hydrogen ionophore II Selectophore™
Synonym(s):
4-Nonadecylpyridine, ETH 1907, Proton ionophore II
Empirical Formula (Hill Notation):
C24H43N
CAS Number:
Molecular Weight:
345.60

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing