Skip to Content
MilliporeSigma
Hydrogen hexachloroiridate(IV) hydrate 99.9% trace metals basis

Hydrogen hexachloroiridate(IV) hydrate

Hydrogen hexachloroiridate(IV) hydrate 99.9% trace metals basis
Synonym(s):
Dihydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Hydrogen iridium hexachloride
Linear Formula:
H2Cl6Ir · xH2O
CAS Number:
Molecular Weight:
406.95 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2