Skip to Content
MilliporeSigma
Hydrocortisone 17-butyrate

Hydrocortisone 17-butyrate

Hydrocortisone 17-butyrate
Synonym(s):
11β,17,21-Trihydroxypregn-4-ene-3,20-dione 17-butyrate, Cortisol 17-butyrate
Empirical Formula (Hill Notation):
C25H36O6
CAS Number:
Molecular Weight:
432.55
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing