Skip to Content
MilliporeSigma
vibrant-m

Hydrocodone Related Compound A

vibrant-m
Synonym(s):
4-Hydroxy-3-methoxy-17-methylmorphinan-6-one, Dihydrothebainone
Empirical Formula (Hill Notation):
C18H23NO3
CAS Number:
Molecular Weight:
301.38

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1