Skip to Content
MilliporeSigma
Hydrazine sulfate salt puriss. p.a., ACS reagent, ≥99.0%

Hydrazine sulfate salt

Hydrazine sulfate salt puriss. p.a., ACS reagent, ≥99.0%
Linear Formula:
NH2NH2 · H2SO4
CAS Number:
Molecular Weight:
130.12
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2