Skip to Content
MilliporeSigma
Hydrazine hydrate solution puriss. p.a., 24-26% in H2O (RT)

Hydrazine hydrate solution

Hydrazine hydrate solution puriss. p.a., 24-26% in H2O (RT)
Synonym(s):
Hydrazinium hydroxide
Linear Formula:
NH2NH2 · H2O
CAS Number:
Molecular Weight:
50.06

Products

Product Category
Formula Weight
Physical Form
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing