Skip to Content
MilliporeSigma
Human Serum NIST® SRM® 909c

Human Serum

Human Serum NIST® SRM® 909c

Products

Product Category
Brand
Biological Source
Manufacturer
Physical Form
Format
Tissue
Chemical Composition
Quality Segment
Geographic Origin
Application
Reference Material Type
Special Grade
Sterilization Method
Technique