![Hispidulin 7-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/237/563/6e6e450a-0509-4e43-8b27-cc06e885a60e/640/6e6e450a-0509-4e43-8b27-cc06e885a60e.png)
Hispidulin 7-glucoside
![Hispidulin 7-glucoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/237/563/6e6e450a-0509-4e43-8b27-cc06e885a60e/640/6e6e450a-0509-4e43-8b27-cc06e885a60e.png)
Synonym(s):
Homoplantaginin, 4′,5,7-Trihydroxy 6-methoxyflavone 7-glucoside, 6-Methoxyapigenin 7-O-β-D-glucoside, 6-Methoxyapigenin 7-O-glucoside, 6-Methylscutellarein 7-glucoside, 7-(β-D-Glucopyranosyloxy)-5-hydroxy-2-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, Dinatin 7-glucoside, Hispidulin 7-β-glucoside, Hispiduloside, Salvitin 7-glucoside, Scutellarein 6-methyl ether 7-glucoside
Empirical Formula (Hill Notation):
C22H22O11
CAS Number:
Molecular Weight:
462.40
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing