Skip to Content
MilliporeSigma
Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate analytical standard

Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate

Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate analytical standard
Synonym(s):
Diethylamino hydroxybenzoyl hexyl benzoate
Empirical Formula (Hill Notation):
C24H31NO4
CAS Number:
Molecular Weight:
397.51
EC Number:

Products

Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing