Skip to Content
MilliporeSigma
Hexanoyl coenzyme A trilithium salt hydrate ≥85%

Hexanoyl coenzyme A trilithium salt hydrate

Hexanoyl coenzyme A trilithium salt hydrate ≥85%
Synonym(s):
Caproyl Coenzyme A trilithium salt
Empirical Formula (Hill Notation):
C27H43Li3N7O17P3S · xH2O
CAS Number:
Molecular Weight:
883.48 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing