Skip to Content
MilliporeSigma
Hexamethyldisilane 98%

Hexamethyldisilane

Hexamethyldisilane 98%
Synonym(s):
Hexamethyldisilane
Linear Formula:
(CH3)3SiSi(CH3)3
CAS Number:
Molecular Weight:
146.38

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing