Skip to Content
MilliporeSigma
Hexafluoroacetone trihydrate 98%

Hexafluoroacetone trihydrate

Hexafluoroacetone trihydrate 98%
Synonym(s):
Hexafluoro-2-propanone trihydrate, Perfluoroacetone trihydrate
Linear Formula:
(CF3)2CO · 3H2O
CAS Number:
Molecular Weight:
220.07
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing