Skip to Content
MilliporeSigma
Hexaflumuron PESTANAL®, analytical standard

Hexaflumuron

Hexaflumuron PESTANAL®, analytical standard
Synonym(s):
N-[3,5-Dichloro-4-(1,1,2,2-tetrafluoroethoxy)phenylcarbamoyl]-2,6-difluorobenzamide
Empirical Formula (Hill Notation):
C16H8Cl2F6N2O3
CAS Number:
Molecular Weight:
461.14

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique