Skip to Content
MilliporeSigma
Hexadecylamine 98%

Hexadecylamine

Hexadecylamine 98%
Synonym(s):
1-Aminohexadecane, Cetylamine, HAD, Hexadecylamine
Linear Formula:
CH3(CH2)15NH2
CAS Number:
Molecular Weight:
241.46

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing