![Hexacosane analytical standard](/deepweb/assets/sigmaaldrich/product/structures/255/197/689ac4ce-2bab-4570-8355-843ccfb5845f/640/689ac4ce-2bab-4570-8355-843ccfb5845f.png)
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing