Skip to Content
MilliporeSigma
Hexaamminecobalt(III) chloride 99%

Hexaamminecobalt(III) chloride

Hexaamminecobalt(III) chloride 99%
Synonym(s):
Cobalt hexammine trichloride, Hexaamminecobalt trichloride
Linear Formula:
[Co(NH3)6]Cl3
CAS Number:
Molecular Weight:
267.48
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing