Skip to Content
MilliporeSigma
Herbimycin A from Streptomyces hygroscopicus ≥95% (HPLC), powder

Herbimycin A from Streptomyces hygroscopicus

Herbimycin A from Streptomyces hygroscopicus ≥95% (HPLC), powder
Empirical Formula (Hill Notation):
C30H42N2O9
CAS Number:
Molecular Weight:
574.66

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing