Skip to Content
MilliporeSigma
Guanosine 3′,5′-cyclic monophosphate sodium salt ≥99% (HPLC), powder

Guanosine 3′,5′-cyclic monophosphate sodium salt

Guanosine 3′,5′-cyclic monophosphate sodium salt ≥99% (HPLC), powder
Synonym(s):
3′,5′-Cyclic GMP monosodium salt, 3′,5′-cGMP-Na, Guanosine 3′,5′-cyclophosphate monosodium salt, cGMP, cyclic GMP, Guanosine 3′,5′-cyclic Monophosphate, Sodium Salt
Empirical Formula (Hill Notation):
C10H11N5O7P · Na
CAS Number:
Molecular Weight:
367.19

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment