
Guanabenz acetate

Synonym(s):
Guanabenz acetate salt, 1-(2,6-Dichlorobenzylideneamino)guanidine acetate salt, WY-8678
Empirical Formula (Hill Notation):
C8H8Cl2N4 · C2H4O2
CAS Number:
Molecular Weight:
291.13
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing