
Granulocyte-Macrophage Colony-Stimulating Factor from rat

Synonym(s):
GM-CSF
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing