Skip to Content
MilliporeSigma
Gram’s safranin solution for microscopy

Gram’s safranin solution

Gram’s safranin solution for microscopy
Synonym(s):
Safranin T solution
Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Chemical Composition
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing