Skip to Content
MilliporeSigma
Gonyautoxin-2-15N7 and Gonyautoxin-3-15N7 solution 10 μg/g in acetic acid (nominal concentration), analytical standard

Gonyautoxin-2-15N7 and Gonyautoxin-3-15N7 solution

Gonyautoxin-2-15N7 and Gonyautoxin-3-15N7 solution 10 μg/g in acetic acid (nominal concentration), analytical standard
Synonym(s):
Toxin GTX2-15N7 and Toxin GTX3-15N7 solution
Empirical Formula (Hill Notation):
C10H1715N7O8S
Molecular Weight:
402.30

Products

Product Category
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade