Skip to Content
MilliporeSigma
Gonyautoxin-1-15N7 and Gonyautoxin-4-15N7 solution 10 μg/g in acetic acid (nominal concentration), analytical standard

Gonyautoxin-1-15N7 and Gonyautoxin-4-15N7 solution

Gonyautoxin-1-15N7 and Gonyautoxin-4-15N7 solution 10 μg/g in acetic acid (nominal concentration), analytical standard
Synonym(s):
Toxin GTX1-15N7 and Toxin GTX4-15N7 solution
Empirical Formula (Hill Notation):
C10H1715N7O9S
Molecular Weight:
418.30

Products

Product Category
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade