Skip to Content
MilliporeSigma
Glyceryl tri(palmitate-d31) endotoxin tested, 98 atom % D, 98% (CP)

Glyceryl tri(palmitate-d31)

Glyceryl tri(palmitate-d31) endotoxin tested, 98 atom % D, 98% (CP)
Synonym(s):
Glyceryl tri(hexadecanoate-d31), Tri(palmitin-d31)
Empirical Formula (Hill Notation):
C51D93H5O6
CAS Number:
Molecular Weight:
900.89

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2