Skip to Content
MilliporeSigma
Glycerol propoxylate-block-ethoxylate average Mn ~5,300

Glycerol propoxylate-block-ethoxylate

Glycerol propoxylate-block-ethoxylate average Mn ~5,300
Empirical Formula (Hill Notation):
C8H22O7
CAS Number:
Molecular Weight:
230.26

Products

Formula Weight
Chemical Composition
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing