Glutamic acid
Synonym(s):
L-Glutamic acid, (S)-2-Aminopentanedioic acid, Glu
Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing