Skip to Content
MilliporeSigma
Glipizide European Pharmacopoeia (EP) Reference Standard

Glipizide

Glipizide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulfonyl}urea, Glipizide
Empirical Formula (Hill Notation):
C21H27N5O4S
CAS Number:
Molecular Weight:
445.54

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique