Skip to Content
MilliporeSigma
Gestrinone ≥97% (HPLC)

Gestrinone

Gestrinone ≥97% (HPLC)
Synonym(s):
(17α)-13-Ethyl-17-hydroxy-18,19-dinorpregna-4,9,11-trien-20-yn-3-one, Ethylnorgestrienone, R-2323, R2323, RU-2323
Empirical Formula (Hill Notation):
C21H24O2
CAS Number:
Molecular Weight:
308.41

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing