Skip to Content
MilliporeSigma
Geranylgeraniol ≥85% (GC)

Geranylgeraniol

Geranylgeraniol ≥85% (GC)
Synonym(s):
all trans-3,7,11-15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol
Empirical Formula (Hill Notation):
C20H34O
CAS Number:
Molecular Weight:
290.48

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing