Skip to Content
MilliporeSigma
Geranyl chloride 95%

Geranyl chloride

Geranyl chloride 95%
Synonym(s):
trans-1-Chloro-3,7-dimethyl-2,6-octadiene
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2Cl
CAS Number:
Molecular Weight:
172.69

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing