Skip to Content
MilliporeSigma
Gefitinib ≥98% (HPLC)

Gefitinib

Gefitinib ≥98% (HPLC)
Synonym(s):
((3-Chloro-4-fluorophenyl)[7-methoxy-6-[3-(morpholin-4-yl)propoxy]quinazolin-4-yl]amine)
Empirical Formula (Hill Notation):
C22H24ClFN4O3
CAS Number:
Molecular Weight:
446.90

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade