Skip to Content
MilliporeSigma
Formycin A from Streptomyces kaniharaensis, ≥98% (HPLC)

Formycin A

Formycin A from Streptomyces kaniharaensis, ≥98% (HPLC)
Synonym(s):
1-C-(7-Amino-1H-pyrazolo[4,3-d]pyrimidin-3-yl)-1,4-anhydro-(1S)-D-Ribitol, 7-Amino-3-(β-D-ribofuranosyl)-1H-pyrazolo[4,3-d]pyrimidine, 8-Aza-9-deazaadenosine, Formycin, NSC 102811
Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing