Skip to Content
MilliporeSigma
Formaldehyde dimethyl acetal for Grignard reactions, ≥99.0% (GC)

Formaldehyde dimethyl acetal

Formaldehyde dimethyl acetal for Grignard reactions, ≥99.0% (GC)
Synonym(s):
Dimethoxymethane, Methylal, Formaldehyde dimethyl acetal
Linear Formula:
CH2(OCH3)2
CAS Number:
Molecular Weight:
76.09

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Special Grade