Skip to Content
MilliporeSigma
Fmoc-Tyr(tBu)-OH ≥98.0% (HPLC)

Fmoc-Tyr(tBu)-OH

Fmoc-Tyr(tBu)-OH ≥98.0% (HPLC)
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-tyrosine, Fmoc-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH
Empirical Formula (Hill Notation):
C28H29NO5
CAS Number:
Molecular Weight:
459.53

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing