Skip to Content
MilliporeSigma
Fmoc-Ser(tBu)-OH ≥98.0% (HPLC)

Fmoc-Ser(tBu)-OH

Fmoc-Ser(tBu)-OH ≥98.0% (HPLC)
Synonym(s):
Fmoc-O-tert-butyl-L-serine, Fmoc-Ser(tBu)-OH
Empirical Formula (Hill Notation):
C22H25NO5
CAS Number:
Molecular Weight:
383.44

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing